Learn More
Hach Company NitriVer 3 TNT Reagent Set, Nitrogen-Nitrite, Low Range
Supplier:  Hach Company 2608345
Encompass
For determination of Nitrite Nitrogen by the NitriVer 3 Diazotization Test 'N Tube™ Method. Hach Method 10019. Range: 0.002 - 0.500 mg/L NO2-N. 50 tests per package.
Catalog No. 50-199-6333