Learn More
Hach Company Zinc Reagent Set, Zincon, 20 mL
Supplier:  Hach Company 2429300
Encompass
For determination of Zinc by the ZincoVer Zincon method. USEPA approved for wastewater analysis (digestion is required for determining total zinc) using Hach Method 8009. Sample Size: 20 mL. Range: 0.01 - 3.00 mg/L Zn. Set contains ZincoVer 5 Powder Pillows and Cyclohexanone for 100 tests. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-5968