Learn More
Cayman Chemical RGyramIde AhydrochLRIde 10mg
SDP
Supplier:  Cayman Chemical 1852810 MG
Encompass_Preferred
A bacterial DNA gyrase inhibitor that disrupts supercoiling activity (IC50 = 3.3 µM); demonstrates antibacterial activity against E. coli, P. aeruginosa, and S. enterica (MICs range from 10-80 µM)
Catalog No. 50-296-7904