Learn More
Abcam Recombinant Human IgG4 Fc Protein, Tag Free (Active), 200UG
Supplier:  Abcam AB181968200UG
Encompass_Preferred
Recombinant Human IgG4 Fc Protein, Tag Free (Active) is a Human Fragment protein, in the 99 to 327 aa range, expressed in HEK 293, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, SPR.
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy constant gamma 4, Ig gamma-4 chain C region, IGHG4
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1677798