Learn More
TARGETMOL CHEMICALS INC RH01386 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T138675MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RH01386 is a small molecule that can prevent ER stress-induced (beta) cell dysfunction and death and inhibits proapoptotic gene expressionhas the potential for type 2 diabetes treatment. RH01386 restores ER stress-impaired glucose-stimulated insulin secretion responses. purity: 99%
Catalog No. 50-258-1323