Learn More
Medchemexpress LLC HY-N1420A 100mg Medchemexpress, Rhamnose (monohydrate) CAS:10030-85-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN1420A100MG
Encompass_Preferred
Medchemexpress, HY-N1420A 100mg Rhamnose (monohydrate) CAS:10030-85-0 Rhamnose monohydrate (L-Rhamnose monohydrate) is a monosaccharide found in plants and bacteria. Rhamnose monohydrate-conjugated immunogens is used in immunotherapies. Rhamnose monohydrate crosses the epithelia via the transcellular pathway and acts as a marker of intestinal absorption. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0336