Learn More
Cayman Chemical RhapontIgenIn 10mg
SDP
Supplier:  Cayman Chemical 1329310 MG
Encompass_Preferred
A natural analog of resveratrol with antioxidant and anti-cancer activity; a mechanism-based, selective inactivator of CYP450 1A1 (IC50 = 400 nM); inhibits the proliferation of cancer cell lines (IC50 = 48 μM)
Catalog No. 50-295-4857