Learn More
Medchemexpress LLC Rhapontigenin | 500-65-2 | 1 ML

Supplier: Medchemexpress LLC HYN222910MM/1ML
Rhapontigenin, a natural analog of resveratrol, exhibits anticancer, antioxidant, antifungal, and antibacterial activities. It acts as a mechanism-based, potent, and selective cytochrome P450 1A1 inactivator with an IC50 of 400 nM, demonstrating high selectivity over P450 1A2 and P450 1B1.
- Anticancer, antioxidant, antifungal, and antibacterial properties
- Potent, selective cytochrome P450 1A1 inactivator (IC50 = 400 nM)
- 400-fold selectivity for P450 1A1 over P450 1A2, 23-fold over P450 1B1
- Molecular weight: 258.27; Formula: C15H14O4; Appearance: Solid, light yellow to yellow; Purity: 99.66%
- Source: Plants (Leguminosae, Trigonella foenum-graecum Linn.)
- Inhibits ADP- and collagen-induced platelet aggregation
- Demonstrates in vivo anti-thrombosis activity
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.