Learn More
Abcam RHC-80267, diacylglycerol lipase inhibitor, 10MG
Supplier:  Abcam AB14175810MG
Encompass_Preferred
MW 394.5 Da, Purity >98%. Selective diacylglycerol lipase inhibitor (IC₅₀ = 4 μM). Enhances acetylcholine induced smooth muscle relaxation in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6440