Learn More
Analytical Sales & Services Rheodyne RheBuild Kit, Rebuild Kit, Ultra High Pressure, Stainless Steel, Unit Count: 12
Supplier:  Analytical Sales & Services 3983
Encompass
The Rheodyne RheBuild Kit is designed for the maintenance of 6-position, 7-port ultra-high pressure switching valves, ensuring precision performance for applications like HPLC. This rebuild kit includes essential components and tools, extending the operational lifespan of the valve and making maintenance straightforward. Its high-quality stainless steel construction and detailed instructions provide reliability in demanding scientific settings.
Catalog No. NC3373364