Learn More
Medchemexpress LLC Rhenium, tricarbonyl(1,10-phenanthroline)[4-(2H-tetrazol-5-yl)pyridinato]-, (OC-6-33)- | 1404104-40-0 | 99.8% | 596.57 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13621310MM/1ML
Encompass_Preferred
Endoplasmic reticulum dye 1 is a promising live cell imaging agent for the detection of exocytotic events at the plasma membrane. It exhibits low cytotoxicity and resistance to photobleaching, making it suitable for imaging both short- and long-time courses.
  • Promising live cell imaging agent
  • Detects exocytotic events at the plasma membrane
  • Exhibits low cytotoxicity
  • Resistant to photobleaching
  • Suitable for imaging short- or long-time courses
Catalog No. 50-003-52341