Learn More
Medchemexpress LLC Rhenium, tricarbonyl(1,10-phenanthroline-κN1,κN10)[4-(2H-tetrazol-5-yl-κN2)pyridinato] | 1404104-40-0 | MFCD00006339 | 99.8% | 596.57 | C21H12N7O3Re | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1362135MG
Encompass_Preferred
Endoplasmic reticulum dye 1 is a rhenium-based fluorescent probe optimized for live-cell imaging of endoplasmic reticulum structures. It displays excitation at 463 nm and emission at 498 nm, and is supplied as a light yellow solid with high purity. Store protected from light and follow recommended solvent storage conditions for best performance.
  • Bright rhenium-based fluorescence for clear endoplasmic reticulum visualization.
  • Excitation 463 nm and emission 498 nm compatible with common fluorescence filter sets.
  • High purity for reproducible imaging results.
  • Light yellow solid that is easy to handle and prepare for experiments.
  • Stable when stored protected from light under recommended conditions.
  • Intended for live-cell imaging applications.
Catalog No. 50-002-21809