Learn More
Waters Corp Waters Performance Maintenance Kit, For 7750E Rheodyne Injector, HPLC
Supplier:  Waters Corp 201000122
Encompass_Preferred
The Waters Performance Maintenance Kit for the 7750E Rheodyne injector contains all the necessary parts to keep your 7750E Rheodyne injector operating at peak performance.
  • Contains all quality parts necessary for peak performance.
  • Includes one rotor.
  • Includes one rotor seal.
  • Includes two allen wrenches.
  • Suitable for HPLC system type.
  • Replacement schedule: as required.
  • One unit per package.
Catalog No. 50-791-553