Learn More
Anton Paar Highly accurate air-bearing motor (minimum torque 100 nNm)For measurements in both rotation and oscillationIdeal for investigation of sample structure
Supplier:  Anton Paar 385253
Encompass_Preferred
MCR 73 SpecificationsBearing design: Air, fine-pored carbonMotor design: Electronically commutated (EC) permanent magnet synchronous motorDisplacement transducer: High-resolution optical encoderNormal force measurement design: n/aMin. torque (rotation): 100 nNmMin. torque (oscillation): 100 nNmMax. torque: 160 mNmTorque resolution: 5 nNmAngular deflection resolution: 8 nradMin. angular velocity: 10-8 rad/sMax. angular velocity / max. speed: 261 rad/s (2,500 rpm)Min. frequency: 10-7 HzMax. frequency: 100 HzNormal force range: n/aNormal force resolution: 0.4 mNAuto Control: YesTruStrain: n/aTruRate: n/a
Catalog No. 50-005-80557