Learn More
Sino Biological Rhesus CD16/FCGR3/Fc gamma RIII Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK9001315
Encompass_Preferred
The Rhesus CD16 / FCGR3 ELISA Pair Set is for the quantitative determination of Rhesus CD16 / FCGR3.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD16 / FCGR3 coated on a 96-well plate. Standards and samples are added to the wells, and any CD16 / FCGR3 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-CD16 / FCGR3 monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of CD16 / FCGR3 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86377