Learn More
Sino Biological Rhesus CD27 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 90049C02H1
Encompass_Preferred
A DNA sequence encoding the rhesus CD27 (XP_001104337.1) (Met1-Ile192) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1142