Learn More
Sino Biological Recombinant Rhesus CD64/FCGR1 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 90017K49HB100
Encompass_Preferred
A DNA sequence encoding the Rhesus FCGR1 (F7BDL2) (Met1-Pro288) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2815