Learn More
Sino Biological Rhesus FCGR3 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90013C08H100
Encompass_Preferred
A DNA sequence encoding the rhesus CD16 (NP_001258584.1) extracellular domain (Met 1-Gly 206) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3499