Learn More
ATCC ATCC Rhesus Rotavirus (MMU 18006) Strain S:USA:79:2 - VR-1739
Supplier:  ATCC VR1739
Encompass
Rhesus rotavirus (MMU 18006, strain S:USA:79:2) is a research isolate intended for propagation and study in permissive cell lines. The culture was isolated from the stool of Macaca mulatta and is supplied for laboratory research under appropriate biosafety conditions.
  • Isolated from the stool of Macaca mulatta with diarrhea.
  • Propagated in MA-104 Clone 1 cells for virus cultivation.
  • Alternate susceptible hosts include BS-C-1, rhesus embryonic monkey kidney cells, and primary cynomolgus monkey kidney cells.
  • Recommended storage: -70°C or colder to preserve viability.
  • Material has been transferred into a different host during processing.
Catalog No. NC3460010