Learn More
American Research Products Inc Rhesus thrombin antithrombin c
Supplier:  American Research Products Inc CSBEQ027237RH
Encompass
Rhesus thrombin-antithrombin compleX (TAT) ELISA Kit from CUSABIO Detection Range 1 25 pg/mL-80 pg/mL Sensitivity 0 312 pg/mL Sample serum plasma Method Competitive
Catalog No. 50-003-27906