Learn More
TARGETMOL CHEMICALS INC RHO-KINASE-IN-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T127215MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rho-Kinase-IN-1 is an inhibitor of ROCK with Kis of 30.5 nM and 3.9 nM for ROCK1 and ROCK2 respectively. Rho-Kinase-IN-1 can be used in studies about excessive cell proliferation remodeling edema and inflammation diseases. purity: 99%
Catalog No. 50-258-1428