Learn More
ABclonal Technology RhoA Rabbit pAb
SDP
Supplier:  ABclonal Technology A139471000UL
Encompass_Preferred
This gene encodes a member of the Rho family of small GTPases, which cycle between inactive GDP-bound and active GTP-bound states and function as molecular switches in signal transduction cascades. Rho proteins promote reorganization of the actin cytoskeleton and regulate cell shape, attachment, and motility. Overexpression of this gene is associated with tumor cell proliferation and metastasis. Multiple alternatively spliced variants have been identified.
Catalog No. 50-001-08518