Learn More
Cayman Chemical RhodamIn B hydrazIde 10mg
SDP
Supplier:  Cayman Chemical 2313310 MG
Encompass_Preferred
A water-soluble fluorescent probe; ex/em spectra = 510/578 nm, respectively; has been used for the detection of copper, peroxynitrite, nitric oxide, hydrogen peroxide, glucose, diacetyl, and hemoglobin
Catalog No. 50-299-4702