Learn More
Enzo Life Sciences Rhodamine 123 (ultra pure) (25mg). CAS: 62669-70-9
SDP
Supplier:  Enzo Life Sciences ENZ52307
Encompass_Preferred
Rhodamine 123 is a cell-permeant, cationic, green-fluorescent dye that is readily sequestered by active mitochondria without cytotoxic effects. Rhodamine 123 can be used in multi-parametric analysis, without fluorescence interference, in combination with common protein labeling dyes such as cyanine-5 and AMCA. With respect to apoptosis, the presence of mitochondrial membrane potential can be probed with rhodamine 123 while the structure and integrity of mitochondria can be assessed using 10-N-nonyl-acridine orange. Wavelength Maxima: Excitation 507nm, Emission 529nm Alternative name: 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl xanthylium chloride. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2633