Learn More
Medchemexpress LLC Rhodamine 6G | 989-38-8 | MFCD00012665 | 98.02% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYD030950MG
Encompass_Preferred
Rhodamine 6G is a membrane-permeable cationic fluorescent probe used to specifically recognize mitochondrial membrane potentials. It attaches to mitochondria, producing bright fluorescence, and exhibits low toxicity to cells at certain concentrations. This makes it a common choice for detecting mitochondria in animal cells, plant cells, and microorganisms.
  • Recognizes mitochondrial membrane potentials
  • Produces bright fluorescence
  • Low toxicity to cells
  • Used for detecting mitochondria
  • Suitable for cell staining in suspension and adherent cells
  • May inhibit tumor growth and reduce metastases
  • Labels leukocytes in the circulatory system
Catalog No. 50-004-10734