Learn More
Santa Cruz Biotechnology Rhodamine Antibody (11H10) 100 µg/ml
Supplier:  Santa Cruz Biotechnology SC57606
Encompass
Rhodamine Antibody (11H10) is a mouse monoclonal IgG1 antibody that detects the Rhodamine protein of non-species reactive by enzyme-linked immunosorbent assay (ELISA). Anti-Rhodamine antibody (11H10) is available as the non-conjugated form, which is crucial for various applications in research and diagnostics. Rhodamine, a member of a family of fluorone dyes, plays a significant role in biological and environmental studies due to its ability to fluoresce, allowing for easy and cost-effective measurement with fluorimeters.
Catalog No. NC3913276