Learn More
Medchemexpress LLC HY-Y0016 100mg Medchemexpress, Rhodamine B CAS:81-88-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0016100MG
Encompass_Preferred
Medchemexpress, HY-Y0016 100mg Rhodamine B CAS:81-88-9 Rhodamine B is a staining fluorescent dye, commonly used for dyeing textiles, paper, soap, leather, and drugs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4667