Learn More
Medchemexpress LLC Rhodamine B | 81-88-9 | MFCD00011931 | >98.0% | 479.02 g/mol | C28H31ClN2O3 | 10mg
SDP
Supplier:  Medchemexpress LLC HYY0016R10MG
Encompass_Preferred
Rhodamine B (Standard) is the analytical standard of Rhodamine B This product is intended for research and analytical applications Rhodamine B is a staining fluorescent dye commonly used for dyeing textiles paper soap leather and agents
Catalog No. 50-002-68930