Learn More
Medchemexpress LLC Rhodamine B hydrazide | 74317-53-6 | MFCD07808762 | 99.9% | 456.58 g/mol | C28H32N4O2 | 50MG

Supplier: Medchemexpress LLC HY12364550MG
Rhodamine B hydrazide is a fluorescent derivative of rhodamine B used as a chemical probe for detection of sulfite, reactive oxygen/nitrogen species, and certain metal ions. It is supplied as a solid research reagent, is soluble in DMSO at 100 mg/mL, and is commonly used in fluorescence-based assays.
- Fluorescent probe with spirocyclic rhodamine B structure.
- Detects sulfite and reactive species such as peroxynitrite and hydroxyl radicals.
- Sensitive to metal ions including copper(II) and mercury(II).
- Molecular formula C28H32N4O2.
- Molecular weight 456.58 g/mol.
- High purity (~99.9%) and supplied in small milligram pack sizes.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.