Learn More
Medchemexpress LLC Rhodamine B isothiocyanate | 36877-69-7 | 97.1% | 536.09 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11269750MG
Encompass_Preferred
Rhodamine B isothiocyanate (RBITC) is a fluorescent dye commonly used in biological imaging and labeling applications. It possesses an isothiocyanate functional group that allows covalent attachment to amino groups on proteins and other biomolecules, facilitating fluorescent labeling of cells and tissues. RBITC emits a bright orange-red fluorescence when excited by green light, making it easily detectable and useful for tracking labeled molecules in complex samples. Its stability and sensitivity have led to its widespread use in various research fields, including cell biology, immunology, and neurobiology.
  • Utilized in biological imaging and labeling
  • Enables covalent attachment to amino groups on proteins and other biomolecules
  • Emits bright orange-red fluorescence under green light excitation
  • Suitable for tracking labeled molecules in complex samples
  • Widely used in cell biology, immunology, and neurobiology research
Catalog No. 50-004-36967