Learn More
AdipoGen Rhodamine B octadecyl est ClO4
SDP
Supplier:  AdipoGen CDXO0022M020
Encompass_Preferred
Chemical. CAS 142179-00-8. Formula C46H67ClN2O7. MW 795.49. Synthetic. Sensitive membrane dye used for potassium fiber-optic sensors and nitrate sensing and other investigations of membranes. Lipophilic energy transfer acceptor from lipophilic fluoresceins in fluorescence energy transfer FRET assays in cell fusion experiments Ex/Em 554/575nm in methanol.
Catalog No. 50-256-0345