Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rhodamine B PEG2-NH2 | 95.3% | 566.13 g/mol | C32H40ClN3O4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1383965MG

Encompass_Preferred

Rhodamine B PEG2-NH2 is a PEG-based PROTAC linker providing a PEG2 spacer with a terminal amine and a Rhodamine B fluorophore for use in PROTAC synthesis and fluorescence-conjugation workflows.

  • PEG-based linker suitable for PROTAC synthesis.
  • Contains Rhodamine B dye for fluorescence labeling.
  • Molecular formula C32H40ClN3O4; molecular weight 566.13 g/mol.
  • High purity of 95.3%.
  • Available in research pack sizes starting at 5 mg.
  • Recommended storage: powder at -20°C for long-term or 4°C for up to two years.

Catalog No. 50-002-23389


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.