Learn More
Jackson Immuno Research Labs RHODAMINE (TRITC) AFFINIPURE
SDP
Supplier:  Jackson Immuno Research Labs 706025148
Encompass_Preferred
AffiniPure F(ab')2 Fragment Donkey Anti-Guinea Pig IgG (H+L) (min X Bov,Ck,Gt,Sy Hms,Hrs,Hu,Ms,Rb,Rat,Shp Sr Prot)
Catalog No. NC1095176