Learn More
Medchemexpress LLC Rhodanine | 141-84-4 | MFCD00005488 | 98.0% | 133.20 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW010365
Encompass_Preferred
Rhodanine is a selective Echovirus 12 proliferation inhibitor and intermediate, playing a role in various biochemical applications. It is utilized in determining tannase activity and serves as a key intermediate in the synthesis of anticancer and antibacterial agents.
  • Acts as a selective echovirus 12 proliferation inhibitor
  • Used to determine tannase activity
  • Can be used in the synthesis of anticancer agents
  • Can be used in the synthesis of antibacterial agents
Catalog No. 50-004-51495