Learn More
Electron Microscopy Sciences Rhodinile Blue Stain Solution 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2616201
Encompass_Preferred
Used in Rhodinile Blue Stain, Stains for hematologic elements. References are Simpson, C.F., Carlisle, J.W., and Mallard, L.: Stain Technology, 45:22 (1970) and Clark, G.: Staining Procedures, Williams and Wilkins Company, Baltimore, 3rd Ed., C. 1973, p. 131.
Catalog No. 50-317-11