Learn More
Cayman Chemical RhodIola rosea RosIn 10mg
SDP
Supplier:  Cayman Chemical 3438310 MG
Encompass_Preferred
A cinnamic glycoside with antidepressant and neurotropic activities; decreases immobility time in the forced swim test in rats at 0.26 mg/kg; reduces chloral hydrate-induced sleep in mice
Catalog No. 50-295-0491