Learn More
Medchemexpress LLC HY-N2425 5mg Medchemexpress, Rhodiosin CAS:86831-54-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN24255MG
Encompass_Preferred
Medchemexpress, HY-N2425 5mg Rhodiosin CAS:86831-54-1 Rhodiosin, isolated from the root of Rhodiola crenulata, is a specific non-competitive cytochrome P450 2D6 inhibitor with an IC50 of 0.420 μM and a Ki of 0.535 μM. Rhodiosin exhibits potent, dose-dependent inhibitory effects on acetylcholinesterase (AChE) with IC50 ranged from 57.50 to 2.43 μg/mL. Rhodiosin exhibits potent DPPH free radical scavenging activities, with an IC50 of 27.77 μM[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0391