Learn More
Strem, An Ascensus Company CAS# 99326-34-8. 100mg. Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, 99%. MFCD00143752
SDP
Supplier:  Strem, An Ascensus Company 450110100MG
Encompass
CAS# 99326-34-8. 100mg. Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, 99%. MFCD00143752. Molecular Weight: 468.34. Molecular Formula: Rh(1,5-C8H12)2+SO3CF3ae. Color/form: dark red xtl. Strem# 45-0110. http://www.strem.com/catalog/v/45-0110/
Catalog No. NC2112549