Learn More
Medchemexpress LLC Rhodium(II) triphenylacetate dimer | 142214-04-8 | MFCD15071082 | >=97.0% | 1355.14 g/mol | C80H60O8Rh2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW033831500MG
Encompass_Preferred
Rhodium(II) triphenylacetate dimer is a dimeric rhodium catalyst widely used in organic synthesis for C-H activation and related metal-catalyzed transformations. Supplied as a solid, it exhibits high selectivity and promotes efficient formation of carbon-carbon and carbon-heteroatom bonds; store protected from light and under inert atmosphere to maintain activity.
  • Dimeric rhodium catalyst for C-H activation and carbene-transfer reactions.
  • High selectivity and reaction rate in catalytic transformations.
  • Solid form with recommended storage to preserve activity.
  • Suitable for method development and scale-up in organic synthesis.
  • Characterized and documented for reproducible results.
Catalog No. 50-002-99412