Learn More
Strem, An Ascensus Company CAS# 14284-92-5. 250mg. Rhodium(III) acetylacetonate, 97+% (99.9%-Rh). MFCD00083144
SDP
Supplier:  Strem, An Ascensus Company 451800250MG
Encompass
CAS# 14284-92-5. 250mg. Rhodium(III) acetylacetonate, 97+% (99.9%-Rh). MFCD00083144. Molecular Weight: 400.24. Molecular Formula: Rh(C5H7O2)3. Color/form: yellow xtl. Strem# 45-1800. http://www.strem.com/catalog/v/45-1800/
Catalog No. 50-901-12213