Learn More
Abcam Recombinant Human Alpha-synuclein protein aggregate (Active), 100UG
Supplier:  Abcam AB218819100UG
Encompass_Preferred
Recombinant Human Alpha-synuclein protein aggregate (Active) is a Human Full Length SNCA protein, in the 1 to 140 aa range with >95% purity, <= 20 EU/mL endotoxin level and suitable for western blot, Functional studies and SDS-PAGE. The predicted molecular weight of ab218819 recombinant protein is 14.5 kDa. - Suitable for Thioflavin T Fluorescence assay - Save time and ensure accurate results - use recombinant Alpha-synuclein (SNCA) protein - Optimal protein bioactivity, stability and reproducibility
Protein/ Peptide/ Enzyme Synonyms: NACP, PARK1, SNCA, Alpha-synuclein, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1468315