Learn More
Akron Biotechnology LLC Recombinant Human Interleukin-2 Lyophilized, GMP Grade, 1 mg vial, AK8223-1000
Supplier:  Akron Biotechnology LLC AK82231000
Encompass
Akron's Recombinant Human Interleukin-2 (rHu IL-2) products are manufactured following all relevant cGMP guidelines for ancillary materials Product Features:Active Substance• Amino acid sequence identical to Proleukin / aldesleukin• Carrier protein-free formulation• E. coli expression system• All raw materials are compliant, controlled, and traceable under Akron’s Quality Management System (QMS)Manufacturing• Type II eCTD MF (#026152) on file with the FDA• Tag-free pharmaceutical processing• Gram-scale production capacityStability• 24-month shelf life• Store at 2-8 °C• Transport with cold packsFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications. This product is not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.
Catalog No. NC1794461