Learn More
Selleck Chemical LLC Rhynchophylline S9400-1MG
SDP
Supplier:  Selleck Chemical LLC S94001MG
Encompass_Preferred
Rhynchophylline (Mitrinermine) an active component isolated from species of the genus Uncaria acts as a calcium channel blocker and is widely used in traditional Chinese medicine mainly for treating ailments of central nervous and cardiovascular systems such as lightheadedness convulsions numbness and hypertension
Catalog No. 50-000-24529