Learn More
Medchemexpress LLC RI-1 | 415713-60-9 | C14H11Cl3N2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1531750MG
Encompass_Preferred
RI-1 is a potent RAD51 inhibitor that acts by covalently binding to cysteine 319 on the RAD51 protein, thereby inactivating it and disrupting homologous recombination in human cells. This action allows RI-1 to sensitize cancer cells to cross-linking chemotherapy and has been shown to reduce triple-negative breast cancer tumor growth in vivo.
  • Inhibits RAD51 by covalently binding to cysteine 319.
  • Disrupts homologous recombination in human cells.
  • Sensitizes human cancer cells to cross-linking chemotherapy.
  • Reduces triple-negative breast cancer tumor growth in mice.
  • Specifically inhibits homologous recombination (HR) in U2OS cells and stimulates single-strand annealing (SSA) in HEK293 cells.
Catalog No. 50-003-76949