Learn More
Medchemexpress LLC 1H-Pyrrole-2,5-dione, 1-(3,4-dichlorophenyl)-3-(4-methoxyphenyl)-4-(4-morpholinyl)- | 1417162-36-7 | 99.9% | 433.28 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY169045MG
Encompass_Preferred
RI-2 is a reversible RAD51 inhibitor that specifically inhibits homologous recombination repair in human cells. It has an IC50 of 44.17 μM for RAD51. RI-2 (150 μM) can significantly sensitize cells.
  • Reversible RAD51 inhibitor
  • Specifically inhibits homologous recombination repair in human cells
  • Induces significant sensitization of cells at 150 μM
Catalog No. 50-208-1914