Learn More
U.S. Pharmacopeia Ribavirin, 36791-04-5, MFCD00058564, 200 mg
Supplier:  U.S. Pharmacopeia 1602706
Encompass
Empirical Formula (Hill Notation): C8H12N4O5, Molecular Weight: 244.20, Synonym: 1-β-D-Ribofuranosyl-1,2,4-triazole-3-carboxamide.
Catalog No. NC0950568