Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ribavirin carboxylic acid | 39925-19-4 | 99.4% | 245.19 g/mol | C8H11N3O6 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1459915MG

Encompass_Preferred

Ribavirin carboxylic acid is the carboxylic acid metabolite of the antiviral ribavirin. Supplied as a high-purity analytical reference, it is intended for metabolite characterization, analytical method development, and antiviral research. The material is a white to off-white solid with defined storage and solubility conditions for laboratory use.

  • Purity 99.42%.
  • Molecular weight 245.19 g/mol.
  • Molecular formula C8H11N3O6.
  • White to off-white solid appearance.
  • Soluble in DMSO at 1 mg/mL with ultrasonic warming.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Includes COA and SDS documentation.

Catalog No. 50-002-21718


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.