Learn More
Medchemexpress LLC Ribavirin carboxylic acid | 39925-19-4 | 99.4% | C8H11N3O6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14599110MG
Encompass_Preferred
Ribavirin carboxylic acid is a metabolite of the antiviral ribavirin and is provided as a solid analytical standard for research use, metabolite identification, and bioanalysis. It has a defined molecular formula and molecular weight and is supplied in small pack sizes suitable for laboratory assays.
  • High purity suitable for analytical and reference applications.
  • Molecular formula C8H11N3O6 and molecular weight 245.19 g/mol.
  • White to off-white solid physical form for easy handling and storage.
  • Intended for metabolite identification and bioanalytical studies.
  • Available in multiple small pack sizes for method development and testing.
Catalog No. 50-002-58604