Learn More
Sigma Aldrich Fine Chemicals Biosciences Ribavirin Related Compound A United States Pharmacopeia (USP) Reference Standard | 39925-19-4 | 15MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 160271715MG
Encompass
Ribavirin Related Compound A United States Pharmacopeia (USP) Reference Standard | Mol Wt: 245.19 | 39925-19-4 | 15MG
Catalog No. 50-186-6592