Learn More
Medchemexpress LLC Ribitol-1-13C (Adonitol-1-13C) | 488-81-3 | 99.5% | 153.14 g/mol | C5H12O5 (1-13C) | 5 MG
SDP
Supplier:  Medchemexpress LLC HY100582S5MG
Encompass_Preferred
Ribitol-1-13C is a carbon-13 labeled ribitol supplied as a high-purity stable isotope reagent for analytical and tracer applications. It is intended for use as an internal standard in metabolomics and isotope-tracing experiments, is highly water-soluble, and requires protection from light and storage under an inert atmosphere for long-term stability.
  • High chemical purity (≈99.5%).
  • Isotopic label at the carbon-1 position (13C) for tracing experiments.
  • Suitable as an internal standard in metabolomics and mass spectrometry.
  • Water-soluble; may require ultrasonic assistance to dissolve at high concentrations.
  • Stable when stored under nitrogen and protected from light; in solvent: -80°C (≈6 months), -20°C (≈1 month).
  • Crystalline powder form, easy to handle for sample preparation.
Catalog No. 50-002-13431